ENA402267673 Enamine

ethyl 2-amino-4-fluoro-3-methylbenzoate

  • MDL number MFCD11519034
  • Cas Number : 1093758-83-8
  • Molecular Weight : 197.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-129645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129645
Availability details for
Quantity
  • Availability
  • Add to Cart