ENA402267674 Enamine

2-chloro-4-(2-chlorophenyl)-1,3-thiazole

  • MDL number MFCD09746497
  • Cas Number : 42444-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-132440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132440
Availability details for
Quantity
  • Availability
  • Add to Cart