ENA402267683 Enamine

5-(propan-2-yl)-4H,5H,6H,7H-[1,3]thiazolo[5,4-c]pyridine-2-carbaldehyde

  • MDL number MFCD17251096
  • Cas Number : 1341653-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132481
Availability details for
Quantity
  • Availability
  • Add to Cart