ENA402267687 Enamine

2-bromo-5-[(2-fluorophenyl)methyl]-1,3,4-thiadiazole

  • MDL number MFCD18345114
  • Cas Number : 1342411-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132485

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132485
Availability details for
Quantity
  • Availability
  • Add to Cart