ENA402267689 Enamine

2-cyclopropylpyridine-3-carboxylic acid

  • MDL number MFCD18255584
  • Cas Number : 1244761-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132487

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132487
Availability details for
Quantity
  • Availability
  • Add to Cart