ENA402267693 Enamine

1-(pyridin-3-ylmethyl)-1H-1,2,3-triazole-4-carbaldehyde

  • MDL number MFCD21715393
  • Cas Number : 915370-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97...99
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132581

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132581
Availability details for
Quantity
  • Availability
  • Add to Cart