ENA402267699 Enamine

5-methyl-1-(pyridin-2-yl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD04111030
  • Cas Number : 340720-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132587
Availability details for
Quantity
  • Availability
  • Add to Cart