ENA402267701 Enamine

6-(2-ethylpiperidin-1-yl)-1,2,3,4-tetrahydroquinoline dihydrochloride

  • MDL number MFCD25371802
  • Cas Number : 1461715-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132589
Availability details for
Quantity
  • Availability
  • Add to Cart