ENA402267705 Enamine

2-(aminomethyl)-1lambda6-thiane-1,1-dione hydrochloride

  • MDL number MFCD25371803
  • Cas Number : 1461707-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132594

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132594
Availability details for
Quantity
  • Availability
  • Add to Cart