ENA402267709 Enamine

2-cyclopropylethanethioamide

  • MDL number MFCD12138805
  • Cas Number : 1154385-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-132605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132605
Availability details for
Quantity
  • Availability
  • Add to Cart