ENA402267713 Enamine

1-(1-bromoethyl)-4-(propan-2-yl)benzene

  • MDL number MFCD11103126
  • Cas Number : 59770-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132611

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132611
Availability details for
Quantity
  • Availability
  • Add to Cart