ENA402267715 Enamine

5-sulfamoyl-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD24497483
  • Cas Number : 1461707-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-132613

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132613
Availability details for
Quantity
  • Availability
  • Add to Cart