ENA402267720 Enamine

2-chloro-1-[3-(trifluoromethyl)phenyl]ethan-1-one

  • MDL number MFCD07779289
  • Cas Number : 439807-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-132620

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132620
Availability details for
Quantity
  • Availability
  • Add to Cart