ENA402267724 Enamine

4-methyl-1-(pyridin-3-yl)-1H-pyrazol-3-amine

  • MDL number MFCD19638977
  • Cas Number : 1339580-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132626
Availability details for
Quantity
  • Availability
  • Add to Cart