ENA402267730 Enamine

ethyl 3-(2-ethoxy-2-oxoethyl)-5-oxopyrrolidine-3-carboxylate

  • MDL number MFCD25371608
  • Cas Number : 91189-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129734
Availability details for
Quantity
  • Availability
  • Add to Cart