ENA402267731 Enamine

5-cyclopropyl-3-methyl-1,2-oxazol-4-amine hydrochloride

  • MDL number MFCD25371811
  • Cas Number : 1461713-39-2
  • Molecular Weight : 174.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-cyclopropyl-3-methylisoxazol-4-amine hydrochloride
Inchi Code 1S/C7H10N2O.ClH/c1-4-6(8)7(10-9-4)5-2-3-5;/h5H,2-3,8H2,1H3;1H
Inchi Key SBPSICGXJAHZTM-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-132657

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-132657
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart