ENA402267732 Enamine

ethyl 2-{[(benzyloxy)carbonyl]amino}-3-sulfamoylpropanoate

  • MDL number MFCD25371609
  • Cas Number : 5466-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129743

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129743
Availability details for
Quantity
  • Availability
  • Add to Cart