ENA402267736 Enamine

3-(1-methyl-1H-1,2,4-triazol-3-yl)aniline

  • MDL number MFCD25371563
  • Cas Number : 1461706-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184...186
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109814

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-109814
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart