ENA402267740 Enamine

1-(2-bromoethyl)-2,3-difluorobenzene

  • MDL number MFCD12025023
  • Cas Number : 126163-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132679

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132679
Availability details for
Quantity
  • Availability
  • Add to Cart