ENA402267742 Enamine

2-chloro-5-(2-chlorophenyl)-1,3,4-thiadiazole

  • MDL number MFCD18344734
  • Cas Number : 36894-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132684

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132684
Availability details for
Quantity
  • Availability
  • Add to Cart