ENA402267762 Enamine

thiane-2-carboxamide

  • MDL number MFCD19212314
  • Cas Number : 18729-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129923

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129923
Availability details for
Quantity
  • Availability
  • Add to Cart