ENA402267771 Enamine

1-ethoxy-3-fluoro-2-nitrobenzene

  • MDL number MFCD13689265
  • Cas Number : 1233952-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-132820

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132820
Availability details for
Quantity
  • Availability
  • Add to Cart