ENA402267779 Enamine

3-(oxolan-2-yl)-1,2-oxazol-5-amine

  • MDL number MFCD24643469
  • Cas Number : 1218915-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68...70
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132850

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132850
Availability details for
Quantity
  • Availability
  • Add to Cart