ENA402267780 Enamine

6-fluoro-1-(2-fluorophenyl)-1H-1,3-benzodiazole-2-thiol

  • MDL number MFCD17967177
  • Cas Number : 1283060-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-132852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132852
Availability details for
Quantity
  • Availability
  • Add to Cart