ENA402267781 Enamine

2-(2-methoxyphenyl)-1H-imidazole-5-carboxylic acid

  • MDL number MFCD07378831
  • Cas Number : 1083318-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132854

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132854
Availability details for
Quantity
  • Availability
  • Add to Cart