ENA402267784 Enamine

2-amino-4-fluoro-5-methylbenzoic acid

  • MDL number MFCD20645600
  • Cas Number : 637347-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129928

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129928
Availability details for
Quantity
  • Availability
  • Add to Cart