ENA402267788 Enamine

3-(1-methyl-1H-1,2,4-triazol-5-yl)aniline

  • MDL number MFCD16851327
  • Cas Number : 1248012-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132864
Availability details for
Quantity
  • Availability
  • Add to Cart