ENA402267789 Enamine

2-{3-[(4-chloro-2-methylphenoxy)methyl]-1H-1,2,4-triazol-5-yl}pyridine

  • MDL number MFCD06297832
  • Cas Number : 620934-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-132865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132865
Availability details for
Quantity
  • Availability
  • Add to Cart