ENA402267791 Enamine

1-cyclohexyl-4-methyl-1H-pyrazol-3-amine

  • MDL number MFCD16845467
  • Cas Number : 1250163-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132869

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132869
Availability details for
Quantity
  • Availability
  • Add to Cart