ENA402267794 Enamine

4-(3-chloro-4-fluorophenoxy)-3-fluorobenzonitrile

  • MDL number MFCD19018400
  • Cas Number : 1410890-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97...99
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132873

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132873
Availability details for
Quantity
  • Availability
  • Add to Cart