ENA402267798 Enamine

4-(oxolan-3-yl)piperidine hydrochloride

  • MDL number MFCD25371829
  • Cas Number : 1461708-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132896

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132896
Availability details for
Quantity
  • Availability
  • Add to Cart