ENA402267809 Enamine

methyl 2-amino-2-(2,4-dichlorophenyl)acetate

  • MDL number MFCD08051825
  • Cas Number : 1008008-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132962

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132962
Availability details for
Quantity
  • Availability
  • Add to Cart