ENA402267810 Enamine

4-{1-[(5-methyl-1,2-oxazol-3-yl)methyl]-1,2,3,6-tetrahydropyridin-4-yl}phenol

  • MDL number MFCD25371830
  • Cas Number : 1311839-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132964

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132964
Availability details for
Quantity
  • Availability
  • Add to Cart