ENA402267812 Enamine

2-(3-nitrophenyl)-1H-imidazole-4-carboxylic acid

  • MDL number MFCD14583645
  • Cas Number : 601494-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184...186
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129985

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129985
Availability details for
Quantity
  • Availability
  • Add to Cart