ENA402267814 Enamine

2-amino-3-(4-chloro-1H-pyrazol-1-yl)propanoic acid hydrochloride

  • MDL number MFCD25371833
  • Cas Number : 1461705-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201...203
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132992

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132992
Availability details for
Quantity
  • Availability
  • Add to Cart