ENA402267838 Enamine

1-(2-amino-1,3-thiazol-4-yl)-2-chloroethan-1-one hydrochloride

  • MDL number MFCD25371835
  • Cas Number : 1461709-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132994

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132994
Availability details for
Quantity
  • Availability
  • Add to Cart