ENA402267862 Enamine

2-(cyclopentylsulfanyl)-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD18923162
  • Cas Number : 1461715-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174...176
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133130

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133130
Availability details for
Quantity
  • Availability
  • Add to Cart