ENA402267864 Enamine

2-(aminomethyl)-2-methylbutanedioic acid hydrochloride

  • MDL number MFCD25371836
  • Cas Number : 86970-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132995
Availability details for
Quantity
  • Availability
  • Add to Cart