ENA402267885 Enamine

2-(chloromethyl)-5-ethyl-1,3,4-thiadiazole

  • MDL number MFCD21288368
  • Cas Number : 1461708-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133293

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133293
Availability details for
Quantity
  • Availability
  • Add to Cart