ENA402267888 Enamine

1-(3-methoxypropanesulfonyl)piperidine

  • MDL number MFCD20360403
  • Cas Number : 1341372-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133320

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133320
Availability details for
Quantity
  • Availability
  • Add to Cart