ENA402267911 Enamine

N-[4-({[2-(3-chlorophenyl)ethyl]amino}methyl)phenyl]acetamide

  • MDL number MFCD13408556
  • Cas Number : 1209424-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133071

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133071
Availability details for
Quantity
  • Availability
  • Add to Cart