ENA402267919 Enamine

3-[(pyridin-3-ylmethyl)amino]propanenitrile

  • MDL number MFCD00047455
  • Cas Number : 33611-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-16261

Safety Information

msds https://www.enaminestore.com/catalog/EN300-16261
Availability details for
Quantity
  • Availability
  • Add to Cart