ENA402268070 Enamine

ethyl({[2-(trifluoromethyl)phenyl]methyl})amine

  • MDL number MFCD09754342
  • Cas Number : 940362-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32246
Availability details for
Quantity
  • Availability
  • Add to Cart