ENA402268072 Enamine

[(3,5-dimethoxyphenyl)methyl](furan-2-ylmethyl)amine

  • MDL number MFCD01135289
  • Cas Number : 418790-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32329

Safety Information

msds https://www.enaminestore.com/catalog/EN300-32329
Availability details for
Quantity
  • Availability
  • Add to Cart