ENA402268085 Enamine

5-(propan-2-yloxy)pyrazine-2-carboxylic acid

  • MDL number MFCD20284630
  • Cas Number : 1344086-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133123

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133123
Availability details for
Quantity
  • Availability
  • Add to Cart