ENA402268095 Enamine

tert-butyl N-(carbamothioylmethyl)carbamate

  • MDL number MFCD09025922
  • Cas Number : 89226-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53880

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-53880
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart