ENA402268101 Enamine

1-(bromomethyl)-2-chloro-4-nitrobenzene

  • MDL number MFCD11847147
  • Cas Number : 42533-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-55
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59267

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P332+P313,P362,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-59267
pictograms GHS05
hstatements H314,H315
Availability details for
Quantity
  • Availability
  • Add to Cart