ENA402268103 Enamine

2-(pyridin-2-yloxy)benzonitrile

  • MDL number MFCD25371866
  • Cas Number : 868755-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72...74
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61737

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61737
Availability details for
Quantity
  • Availability
  • Add to Cart