ENA402268106 Enamine

2-amino-5-bromo-4-fluorobenzoic acid

  • MDL number MFCD06409112
  • Cas Number : 143945-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65910

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-65910
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart