ENA402268108 Enamine

1-methyl-1,2,3,4-tetrahydroquinoline-3-carboxylic acid

  • MDL number MFCD20647292
  • Cas Number : 72594-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133124
Availability details for
Quantity
  • Availability
  • Add to Cart